accessibility menu, dialog, popup

UserName

CH3NNOONHH3CCH3NONH3COOHNFK

CAS RN 871038-72-1

IUPAC Name: potassium 4-{[(4-fluorophenyl)methyl]carbamoyl}-1-methyl-2-{2-[(5-methyl-1,3,4-oxadiazol-2-yl)formamido]propan-2-yl}-6-oxo-1,6-dihydropyrimidin-5-olate
Synonyms: raltegravir
Molecular Weight (g/mol): 482.51
Molecular Formula: C20H20FKN6O5
InChi Key: IFUKBHBISRAZTF-UHFFFAOYSA-M
SMILES: [K+].CN1C(=O)C([O-])=C(N=C1C(C)(C)NC(=O)C1=NN=C(C)O1)C(=O)NCC1=CC=C(F)C=C1
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results