accessibility menu, dialog, popup

UserName

CAS RN 863918-78-9

H3COHOOH(E/Z)(E/Z)

CAS RN 863918-78-9

IUPAC Name: 4-(2-{4-[2-(4-hydroxyphenyl)ethenyl]-3-methoxyphenyl}ethenyl)phenol
Synonyms: 4-(2-{4-[2-(4-hydroxyphenyl)ethenyl]-3-methoxyphenyl}ethenyl)phenol
Molecular Weight (g/mol): 344.41
Molecular Formula: C23H20O3
InChi Key: FGYNZFHVGOFCMD-UHFFFAOYSA-N
SMILES: COC1=C(C=CC2=CC=C(O)C=C2)C=CC(C=CC2=CC=C(O)C=C2)=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results