accessibility menu, dialog, popup

UserName

CAS RN 850567-24-7

BOHNOHOH

CAS RN 850567-24-7

IUPAC Name: [3-(cyclopentylcarbamoyl)phenyl]boronic acid
Synonyms: 3-(cyclopentylcarbamoyl)phenylboronic acid
Molecular Weight (g/mol): 233.07
Molecular Formula: C12H16BNO3
InChi Key: KIOBFDGZJVJNBP-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=CC=C1)C(=O)NC1CCCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results