CAS RN 82768-85-2
CAS RN 82768-85-2
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
(3S)-2-[(2S)-2-[[(1S)-1-Carboxy-3-phenylpropyl]amino]propanoyl]-1,2,3,4-tetrahydroisoquinoline-3-carboxylic Acid (Quinaprilat), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.