accessibility menu, dialog, popup

UserName

ClH3COO(S)NH(S)H3CON(S)HOOH

CAS RN 82586-55-8

IUPAC Name: hydrogen (3S)-2-[(2S)-2-{[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino}propanoyl]-1,2,3,4-tetrahydroisoquinoline-3-carboxylic acid chloride
Synonyms: hydrogen quinapril chloride
Molecular Weight (g/mol): 474.98
Molecular Formula: C25H31ClN2O5
InChi Key: IBBLRJGOOANPTQ-JKVLGAQCSA-N
SMILES: [H+].[Cl-].CCOC(=O)[C@H](CCC1=CC=CC=C1)N[C@@H](C)C(=O)N1CC2=CC=CC=C2C[C@H]1C(O)=O
Molecular Weight (g/mol) 
  • (5)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (3)
  • (3)
  • (5)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)