CAS RN 82297-89-0
CAS RN 82297-89-0
IUPAC Name:
magnesium(2+) 4-fluoro-3-methylbenzen-1-ide bromide
Synonyms:
magnesium(2+) 4-fluoro-3-methylbenzen-1-ide bromide
Molecular Weight (g/mol):
213.33
Molecular Formula:
C7H6BrFMg
InChi Key:
RDVDTHIVHSZVCT-UHFFFAOYSA-M
SMILES:
[Mg++].[Br-].CC1=C[C-]=CC=C1F