accessibility menu, dialog, popup

UserName

CAS RN 822-21-9

H3COH

CAS RN 822-21-9

IUPAC Name: hexadec-7-yn-1-ol
Synonyms: hexadec-7-yn-1-ol
Molecular Weight (g/mol): 238.42
Molecular Formula: C16H30O
InChi Key: UFLJPANUONDNPT-UHFFFAOYSA-N
SMILES: CCCCCCCCC#CCCCCCCO

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results