CAS RN 82104-74-3
CAS RN 82104-74-3
IUPAC Name:
1-oxo-1,3-dihydro-2-benzofuran-5-carbonitrile
Synonyms:
1-oxo-3H-2-benzofuran-5-carbonitrile
Molecular Weight (g/mol):
159.14
Molecular Formula:
C9H5NO2
InChi Key:
XEEGWTLAFIZLSF-UHFFFAOYSA-N
SMILES:
O=C1OCC2=CC(=CC=C12)C#N
1
–
1
of
1
results
1
5-Cyanophthalide, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.