CAS RN 82-28-0
CAS RN 82-28-0
IUPAC Name:
1-amino-2-methyl-9,10-dihydroanthracene-9,10-dione
Synonyms:
1-amino-2-methylanthracene-9,10-dione
Molecular Weight (g/mol):
237.26
Molecular Formula:
C15H11NO2
InChi Key:
ZLCUIOWQYBYEBG-UHFFFAOYSA-N
SMILES:
CC1=CC=C2C(=O)C3=CC=CC=C3C(=O)C2=C1N
1
–
1
of
1
results
1
Disperse Orange 11 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality