CAS RN 81403-68-1
IUPAC Name:
hydrogen N-{3-[(4-amino-6,7-dimethoxyquinazolin-2-yl)(methyl)amino]propyl}oxolane-2-carboxamide chloride
Synonyms:
uroxatral alfuzosin hydrochloride
Molecular Weight (g/mol):
425.91
Molecular Formula:
C19H28ClN5O4
InChi Key:
YTNKWDJILNVLGX-UHFFFAOYNA-N
SMILES:
[H+].[Cl-].COC1=C(OC)C=C2C(N)=NC(=NC2=C1)N(C)CCCNC(=O)C1CCCO1
1
–
3
of
3
results
1
Alfuzosin Hydrochloride, TRC
CAS: 81403-68-1 Molecular Formula: C19 H27 N5 O4 . Cl H Molecular Weight (g/mol): 425.91 Synonym: Alfuzosin hydrochloride,(2RS)-N-[3-[(4-Amino-6,7-dimethoxyquinazolin-2-yl)methylamino]propyl]tetrahydrofuran-2-carboxamide hydrochloride,SL 77-499-10,SL 77499-10,Urion,Uroxatral,Xatral,Alfoten IUPAC Name: N-[3-[(4-amino-6,7-dimethoxyquinazolin-2-yl)-methylamino]propyl]oxolane-2-carboxamide;hydrochloride SMILES: Cl.COc1cc2nc(nc(N)c2cc1OC)N(C)CCCNC(=O)C3CCCO3
2
Alfuzosin Hydrochloride, Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.