accessibility menu, dialog, popup

UserName

CAS RN 81239-45-4

IUPAC Name: 3-(benzyldimethylazaniumyl)propane-1-sulfonate
Synonyms: 3-(benzyldimethylammonio)propane-1-sulfonate
Molecular Weight (g/mol): 257.35
Molecular Formula: C12H19NO3S
InChi Key: MEJASPJNLSQOAG-UHFFFAOYSA-N
SMILES: C[N+](C)(CCCS([O-])(=O)=O)CC1=CC=CC=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results