accessibility menu, dialog, popup

UserName

CAS RN 81110-73-8

H3COSOHNOO

CAS RN 81110-73-8

IUPAC Name: benzyl 2-{2-[(acetylsulfanyl)methyl]-3-phenylpropanamido}acetate
Synonyms: racecadotril
Molecular Weight (g/mol): 385.48
Molecular Formula: C21H23NO4S
InChi Key: ODUOJXZPIYUATO-UHFFFAOYNA-N
SMILES: CC(=O)SCC(CC1=CC=CC=C1)C(=O)NCC(=O)OCC1=CC=CC=C1
Certifications/Compliance 
  • (1)
  • (1)
Industry Type 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results