accessibility menu, dialog, popup

UserName

CAS RN 81103-79-9

2H2H2H2H2H2H2H2H2H2H

CAS RN 81103-79-9

IUPAC Name: (²H₁₀)-9H-fluorene
Synonyms: (²H₁₀)fluorene
Molecular Weight (g/mol): 176.28
Molecular Formula: C13H10
InChi Key: NIHNNTQXNPWCJQ-QNNBDYQESA-N
SMILES: [2H]C1=C([2H])C([2H])=C2C(=C1[2H])C1=C([2H])C([2H])=C([2H])C([2H])=C1C2([2H])[2H]
Molecular Weight (g/mol) 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Percent Purity 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)