accessibility menu, dialog, popup

UserName

CH3(R)OO(R)CH3(S)O(R)(R)H3COH3C(S)OH(S)CH3O(S)CH3(R)O(S)O(R)CH3(S)(R)HONCH3H3C(R)CH3(R)H3CO(R)CH3(R)HO(S)H3COHOH3C

CAS RN 81103-11-9

IUPAC Name: (3R,4S,5S,6R,7R,9R,11R,12R,13S,14R)-6-{[(2S,3R,4S,6R)-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl]oxy}-14-ethyl-12,13-dihydroxy-4-{[(2R,4R,5S,6S)-5-hydroxy-4-methoxy-4,6-dimethyloxan-2-yl]oxy}-7-methoxy-3,5,7,9,11,13-hexamethyl-1-oxacyclotetradecane-2,10-dione
Synonyms: clarithromycin clarithromycinum 6-O-methylerythromycin A 克拉霉素 clarithromycina 6-O-methylerythromycin
Molecular Weight (g/mol): 747.96
Molecular Formula: C38H69NO13
InChi Key: AGOYDEPGAOXOCK-KCBOHYOISA-N
SMILES: CC[C@H]1OC(=O)[C@H](C)[C@@H](O[C@H]2C[C@@](C)(OC)[C@@H](O)[C@H](C)O2)[C@H](C)[C@@H](O[C@@H]2O[C@H](C)C[C@@H]([C@H]2O)N(C)C)[C@@](C)(C[C@@H](C)C(=O)[C@H](C)[C@@H](O)[C@]1(C)O)OC
Molecular Weight (g/mol) 
  • (6)
  • (2)
  • (3)
Quantity 
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
Percent Purity 
  • (5)
  • (1)
Color 
  • (2)
  • (3)
Physical Form 
  • (2)
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (2)
  • (6)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (5)
  • (1)
  • (2)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)