CAS RN 81029-05-2
CAS RN 81029-05-2
IUPAC Name:
7,9-diamino-N,N-diethyl-6-methyl-5a,9a-dihydro-3H-phenoxazin-3-iminium chloride
Synonyms:
brilliant cresyl blue
Molecular Weight (g/mol):
334.85
Molecular Formula:
C17H23ClN4O
InChi Key:
SHMCAAFKAHAURK-UHFFFAOYNA-M
SMILES:
[Cl-].CC[N+](CC)=C1C=CC2=NC3C(OC2=C1)C(C)=C(N)C=C3N
1
–
1
of
1
results
1
Promotions Available
Thermo Scientific Chemicals Brilliant cresyl Blue, pure, high purity biological stain
Thermo Scientific Chemicals Brilliant cresyl Blue, pure, high purity biological stain
CAS: 81029-05-2 Molecular Formula: 0·5 ZnCl4 MDL Number: MFCD00148902