accessibility menu, dialog, popup

UserName

CAS RN 80841-78-7

CH3OOOCl

CAS RN 80841-78-7

IUPAC Name: 4-(chloromethyl)-5-methyl-2H-1,3-dioxol-2-one
Synonyms: 4-(chloromethyl)-5-methyl-1,3-dioxol-2-one
Molecular Weight (g/mol): 148.54
Molecular Formula: C5H5ClO3
InChi Key: QCLFSYYUWPUWQR-UHFFFAOYSA-N
SMILES: CC1=C(CCl)OC(=O)O1
Molecular Weight (g/mol) 
  • (1)
  • (3)
Boiling Point 
  • (3)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results