accessibility menu, dialog, popup

UserName

CAS RN 8025-81-8

CH3O(S)(R)OHOO(R)CH3(R)OCH3ONH3CCH3(R)H3C(R)O(S)OOH3COCH3HOHOH3COOHNCH3H3C(E)(E)

CAS RN 8025-81-8

IUPAC Name: 2-[(4R,5S,6S,7R,9R,10R,11E,13E,16R)-6-({5-[(4,5-dihydroxy-4,6-dimethyloxan-2-yl)oxy]-4-(dimethylamino)-3-hydroxy-6-methyloxan-2-yl}oxy)-10-{[5-(dimethylamino)-6-methyloxan-2-yl]oxy}-4-hydroxy-5-methoxy-9,16-dimethyl-2-oxo-1-oxacyclohexadeca-11,13-dien-7-yl]acetaldehyde
Synonyms: spiramycin
Molecular Weight (g/mol): 843.07
Molecular Formula: C43H74N2O14
InChi Key: ACTOXUHEUCPTEW-JMRHEKERSA-N
SMILES: CO[C@H]1[C@H](O)CC(=O)O[C@H](C)C\C=C\C=C\[C@H](OC2CCC(C(C)O2)N(C)C)[C@H](C)C[C@H](CC=O)[C@@H]1OC1OC(C)C(OC2CC(C)(O)C(O)C(C)O2)C(C1O)N(C)C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results