CAS RN 8007-47-4
CAS RN 8007-47-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
4
of
4
results
1
Canada balsam, natural filtered
CAS: 8007-47-4 MDL Number: MFCD00132800 Synonym: Balsam Canada
2
Canada Balsam, Technical Grade, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
3
Canada Balsam, Technical Grade, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
4
Canada Balsam, Technical Grade, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.