accessibility menu, dialog, popup

UserName

NNOOSOOOONH2NSOOOHNNNaNa(E)(E)

CAS RN 8005-03-6

IUPAC Name: disodium (3E)-5-amino-6-[(1E)-2-(4-nitrophenyl)diazen-1-yl]-4-oxo-3-(2-phenylhydrazin-1-ylidene)-3,4-dihydronaphthalene-2,7-disulfonate
Synonyms: disodium (6E)-4-amino-3-[(1E)-2-(4-nitrophenyl)diazen-1-yl]-5-oxo-6-(2-phenylhydrazin-1-ylidene)naphthalene-2,7-disulfonate
Molecular Weight (g/mol): 616.49
Molecular Formula: C22H14N6Na2O9S2
InChi Key: HKBVRFLHNUEVRO-HMDKGIQMSA-L
SMILES: [Na+].[Na+].NC1=C2C(=O)\C(=N/NC3=CC=CC=C3)C(=CC2=CC(=C1\N=N\C1=CC=C(C=C1)[N+]([O-])=O)S([O-])(=O)=O)S([O-])(=O)=O
Quantity 
  • (3)
  • (2)
  • (1)
  • (1)
Color 
  • (2)
  • (2)
Grade 
  • (4)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (4)
  • (2)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)
  • (4)