CAS RN 80-92-2
CAS RN 80-92-2
IUPAC Name:
(1S,3aS,3bR,5aR,7R,9aS,9bS,11aS)-1-[(1S)-1-hydroxyethyl]-9a,11a-dimethyl-hexadecahydro-1H-cyclopenta[a]phenanthren-7-ol
Synonyms:
pregnanediol
Molecular Weight (g/mol):
320.52
Molecular Formula:
C21H36O2
InChi Key:
YWYQTGBBEZQBGO-BERLURQNSA-N
SMILES:
C[C@H](O)[C@H]1CC[C@H]2[C@@H]3CC[C@@H]4C[C@H](O)CC[C@]4(C)[C@H]3CC[C@]12C
1
–
2
of
2
results
1
Pregnanediol (5beta-Pregnane-3alpha,20(S)-diol), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
Pregnanediol (5beta-Pregnane-3alpha,20(S)-diol), LoGiCal
LoGiCal delivers certified reference materials you can trust as neats, native solutions and stable isotope labelled solutions, with precise documentation and expert support. From forensic to clinical labs, choose LoGiCal for confidence, compliance, and analytical excellence.