CAS RN 79973-42-5
CAS RN 79973-42-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Bis(pentamethylcyclopentadienyl)cobalt(III) hexafluorophosphate, 98%
CAS: 79973-42-5 Molecular Formula: C20H30CoF6P MDL Number: MFCD00269818