accessibility menu, dialog, popup

UserName

CAS RN 796600-10-7

NHONCl

CAS RN 796600-10-7

IUPAC Name: 2-chloro-4-(piperidin-4-yloxy)benzonitrile
Synonyms: 2-chloro-4-(piperidin-4-yloxy)benzonitrile
Molecular Weight (g/mol): 236.7
Molecular Formula: C12H13ClN2O
InChi Key: AZTMLAAAPWXHMV-UHFFFAOYSA-N
SMILES: ClC1=C(C=CC(OC2CCNCC2)=C1)C#N

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results