accessibility menu, dialog, popup

UserName

CAS RN 79660-72-3

H3CNNFONFFOHO

CAS RN 79660-72-3

IUPAC Name: 6,8-difluoro-1-(2-fluoroethyl)-7-(4-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid
Synonyms: fleroxacin quinodis FLRX fleroxicin 氟罗沙星 megalone grünenthal brand of fleroxacin fleroxacin (substance) fleroxacino fleroxacine
Molecular Weight (g/mol): 369.34
Molecular Formula: C17H18F3N3O3
InChi Key: XBJBPGROQZJDOJ-UHFFFAOYSA-N
SMILES: CN1CCN(CC1)C1=C(F)C=C2C(=O)C(=CN(CCF)C2=C1F)C(O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results