CAS RN 79538-32-2
CAS RN 79538-32-2
IUPAC Name:
(2,3,5,6-tetrafluoro-4-methylphenyl)methyl 3-[(1Z)-2-chloro-3,3,3-trifluoroprop-1-en-1-yl]-2,2-dimethylcyclopropane-1-carboxylate
Synonyms:
forza tetrafluthrin epa pesticide chemical code 128912 tefluthrine
Molecular Weight (g/mol):
418.74
Molecular Formula:
C17H14ClF7O2
InChi Key:
ZFHGXWPMULPQSE-WTKPLQERNA-N
SMILES:
CC1=C(F)C(F)=C(COC(=O)C2C(\C=C(/Cl)C(F)(F)F)C2(C)C)C(F)=C1F