accessibility menu, dialog, popup

UserName

CAS RN 79424-03-6

CH3OOFFF

CAS RN 79424-03-6

IUPAC Name: ethyl 4,4,4-trifluorobut-2-ynoate
Synonyms: ethyl 4,4,4-trifluorobut-2-ynoate
Molecular Weight (g/mol): 166.1
Molecular Formula: C6H5F3O2
InChi Key: SFDRHPQGYUYYNX-UHFFFAOYSA-N
SMILES: CCOC(=O)C#CC(F)(F)F
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (1)