accessibility menu, dialog, popup

UserName

CAS RN 79127-80-3

H3COONHOO

CAS RN 79127-80-3

IUPAC Name: ethyl N-[2-(4-phenoxyphenoxy)ethyl]carbamate
Synonyms: fenoxycarb insegar
Molecular Weight (g/mol): 301.34
Molecular Formula: C17H19NO4
InChi Key: HJUFTIJOISQSKQ-UHFFFAOYSA-N
SMILES: CCOC(=O)NCCOC1=CC=C(OC2=CC=CC=C2)C=C1
Concentration 
  • (1)
  • (1)
Solvent or Matrix 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results