CAS RN 79069-50-4
CAS RN 79069-50-4
IUPAC Name:
tert-butyl N-(1-oxopropan-2-yl)carbamate
Synonyms:
tert-butyl N-(1-oxopropan-2-yl)carbamate
Molecular Weight (g/mol):
173.21
Molecular Formula:
C8H15NO3
InChi Key:
OEQRZPWMXXJEKU-UHFFFAOYNA-N
SMILES:
CC(NC(=O)OC(C)(C)C)C=O