accessibility menu, dialog, popup

UserName

CH3NHCH3CH3H3CCH3

CAS RN 79-55-0

IUPAC Name: 1,2,2,6,6-pentamethylpiperidin-1-ium
Synonyms: pempidine
Molecular Weight (g/mol): 156.29
Molecular Formula: C10H22N
InChi Key: XULIXFLCVXWHRF-UHFFFAOYSA-O
SMILES: C[NH+]1C(C)(C)CCCC1(C)C
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (3)
  • (1)
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (3)
  • (4)
  • (2)
  • (1)
  • (1)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)