accessibility menu, dialog, popup

UserName

CAS RN 78686-77-8

CH3OONClBr

CAS RN 78686-77-8

IUPAC Name: methyl 5-bromo-6-chloropyridine-3-carboxylate
Synonyms: methyl 5-bromo-6-chloropyridine-3-carboxylate
Molecular Weight (g/mol): 250.48
Molecular Formula: C7H5BrClNO2
InChi Key: WINGWVOUOFMOJQ-UHFFFAOYSA-N
SMILES: COC(=O)C1=CN=C(Cl)C(Br)=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results