accessibility menu, dialog, popup

UserName

CAS RN 78191-00-1

CH3ONCH3CH3O

CAS RN 78191-00-1

IUPAC Name: N-methoxy-N-methylacetamide
Synonyms: N-methoxy-N-methylacetamide
Molecular Weight (g/mol): 103.12
Molecular Formula: C4H9NO2
InChi Key: OYVXVLSZQHSNDK-UHFFFAOYSA-N
SMILES: CON(C)C(C)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results