accessibility menu, dialog, popup

UserName

CAS RN 78-88-6

ClClH2C

CAS RN 78-88-6

IUPAC Name: 2,3-dichloroprop-1-ene
Synonyms: 2,3-dichloropropene
Molecular Weight (g/mol): 110.97
Molecular Formula: C3H4Cl2
InChi Key: FALCMQXTWHPRIH-UHFFFAOYSA-N
SMILES: ClCC(Cl)=C
Molecular Weight (g/mol) 
  • (4)
  • (5)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)
  • (3)
  • (3)
  • (4)
  • (5)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (2)
  • (1)