accessibility menu, dialog, popup

UserName

CAS RN 78-34-2

CH3OPSOCH3SOOSPSOH3COH3C

CAS RN 78-34-2

IUPAC Name: O,O-diethyl [(3-{[diethoxy(sulfanylidene)-λ⁵-phosphanyl]sulfanyl}-1,4-dioxan-2-yl)sulfanyl]phosphonothioate
Synonyms: dioxathion 敌f磷
Molecular Weight (g/mol): 456.52
Molecular Formula: C12H26O6P2S4
InChi Key: VBKKVDGJXVOLNE-UHFFFAOYNA-N
SMILES: CCOP(=S)(OCC)SC1OCCOC1SP(=S)(OCC)OCC
Molecular Weight (g/mol) 
  • (1)
  • (1)
Solvent or Matrix 
  • (1)
  • (1)
Concentration 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results