accessibility menu, dialog, popup

UserName

CAS RN 78-26-2

CH3CH3HOHO

CAS RN 78-26-2

IUPAC Name: 2-methyl-2-propylpropane-1,3-diol
Synonyms: 2-methyl-2-propylpropane-1,3-diol
Molecular Weight (g/mol): 132.2
Molecular Formula: C7H16O2
InChi Key: JVZZUPJFERSVRN-UHFFFAOYSA-N
SMILES: CCCC(C)(CO)CO
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results