CAS RN 7787-61-3
CAS RN 7787-61-3
IUPAC Name:
bismuth(3+) trifluoride
Synonyms:
bismuth trifluoride
Molecular Weight (g/mol):
265.98
Molecular Formula:
BiF3
InChi Key:
BRCWHGIUHLWZBK-UHFFFAOYSA-K
SMILES:
[F-].[F-].[F-].[Bi+3]
1
–
1
of
1
results
1
Bismuth(III) fluoride, anhydrous, 99%
CAS: 7787-61-3 Molecular Formula: BiF3 Molecular Weight (g/mol): 265.976 MDL Number: MFCD00010893 InChI Key: BRCWHGIUHLWZBK-UHFFFAOYSA-K Synonym: bismuth fluoride,bismuth iii fluoride,bismuthine, trifluoro,trifluorobismuthine,bismuth trifluoride,bismuth iii trifluoride,acmc-1bd9n,ksc378g8f PubChem CID: 82233 IUPAC Name: trifluorobismuthane SMILES: F[Bi](F)F