accessibility menu, dialog, popup

UserName

CAS RN 7787-20-4

H3CH3C(S)(R)CH3O

CAS RN 7787-20-4

IUPAC Name: (1R,4S)-1,3,3-trimethylbicyclo[2.2.1]heptan-2-one
Synonyms: fenchone, (+-)- fenchone, (1r)- fenchone, (1s)- 1,3,3-trimethylnorcamphor trimethylnorbornan-2-one (+-)-fenchone fenchone, (1R)- fenchone, (1S)- fenchon d-fenchone (natural)
Molecular Weight (g/mol): 152.24
Molecular Formula: C10H16O
InChi Key: LHXDLQBQYFFVNW-OIBJUYFYSA-N
SMILES: CC1(C)[C@H]2CC[C@](C)(C2)C1=O
Boiling Point 
  • (4)
Molecular Weight (g/mol) 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (2)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)