accessibility menu, dialog, popup

UserName

CAS RN 7784-18-1

FFFAl

CAS RN 7784-18-1

IUPAC Name: aluminium(3+) trifluoride
Synonyms: aluminium fluoride
Molecular Weight (g/mol): 83.98
Molecular Formula: AlF3
InChi Key: KLZUFWVZNOTSEM-UHFFFAOYSA-K
SMILES: [F-].[F-].[F-].[Al+3]
Percent Purity 
  • (3)
  • (3)
  • (3)
Physical Form 
  • (6)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (9)
  • (3)
  • (3)
  • (3)
  • (6)
  • (3)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)