accessibility menu, dialog, popup

UserName

CAS RN 7783-63-3

FFFFTi

CAS RN 7783-63-3

IUPAC Name: titanium(4+) tetrafluoride
Synonyms: titanium tetrafluoride
Molecular Weight (g/mol): 123.86
Molecular Formula: F4Ti
InChi Key: XROWMBWRMNHXMF-UHFFFAOYSA-J
SMILES: [F-].[F-].[F-].[F-].[Ti+4]
Physical Form 
  • (2)
  • (2)
Quantity 
  • (1)
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (2)
  • (1)