accessibility menu, dialog, popup

UserName

CAS RN 776-41-0

H3COOHN

CAS RN 776-41-0

IUPAC Name: ethyl 1H-indole-3-carboxylate
Synonyms: INDOLE3carboxylicacidethylester
Molecular Weight (g/mol): 189.21
Molecular Formula: C11H11NO2
InChi Key: XOUHVMVYFOXTMN-UHFFFAOYSA-N
SMILES: CCOC(=O)C1=CNC2=CC=CC=C12

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results