accessibility menu, dialog, popup

UserName

CAS RN 7755-92-2

ONNH2

CAS RN 7755-92-2

IUPAC Name: 4-formylpiperazin-1-ium
Synonyms: 4-formylpiperazin-1-ium
Molecular Weight (g/mol): 115.16
Molecular Formula: C5H11N2O
InChi Key: MSSDTZLYNMFTKN-UHFFFAOYSA-O
SMILES: O=CN1CC[NH2+]CC1
Molecular Weight (g/mol) 
  • (3)
  • (5)
Quantity 
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
Boiling Point 
  • (2)
  • (3)
Percent Purity 
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (3)
  • (3)
  • (3)
  • (5)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (2)
  • (3)
  • (3)
  • (2)
  • (2)