accessibility menu, dialog, popup

UserName

CAS RN 775-00-8

NH3FFF

CAS RN 775-00-8

IUPAC Name: 2-[4-(trifluoromethyl)phenyl]ethan-1-aminium
Synonyms: 2-[4-(trifluoromethyl)phenyl]ethanaminium
Molecular Weight (g/mol): 190.19
Molecular Formula: C9H11F3N
InChi Key: HMFOBPNVAAAACP-UHFFFAOYSA-O
SMILES: [NH3+]CCC1=CC=C(C=C1)C(F)(F)F
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results