accessibility menu, dialog, popup

UserName

CAS RN 773871-39-9

H3COOHFFF

CAS RN 773871-39-9

IUPAC Name: [4-methoxy-2-(trifluoromethyl)phenyl]methanol
Synonyms: [4-methoxy-2-(trifluoromethyl)phenyl]methanol
Molecular Weight (g/mol): 206.16
Molecular Formula: C9H9F3O2
InChi Key: MEQLJIHKRJZDPW-UHFFFAOYSA-N
SMILES: COC1=CC(=C(CO)C=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results