accessibility menu, dialog, popup

UserName

CAS RN 7730-20-3

NHONH2OHF

CAS RN 7730-20-3

IUPAC Name: (2S)-2-azaniumyl-3-(6-fluoro-1H-indol-3-yl)propanoate
Synonyms: (2S)-2-ammonio-3-(6-fluoro-1H-indol-3-yl)propanoate
Molecular Weight (g/mol): 222.22
Molecular Formula: C11H11FN2O2
InChi Key: YMEXGEAJNZRQEH-VIFPVBQESA-N
SMILES: [NH3+][C@@H](CC1=CNC2=CC(F)=CC=C12)C([O-])=O
Molecular Weight (g/mol) 
  • (1)
  • (2)
Quantity 
  • (2)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results