accessibility menu, dialog, popup

UserName

CAS RN 771581-99-8

H3COFFFNH2

CAS RN 771581-99-8

IUPAC Name: 1-[2-methoxy-4-(trifluoromethyl)phenyl]methanamine
Synonyms: 1-[2-methoxy-4-(trifluoromethyl)phenyl]methanamine
Molecular Weight (g/mol): 205.18
Molecular Formula: C9H10F3NO
InChi Key: FEOVMOIWRLRBFP-UHFFFAOYSA-N
SMILES: COC1=C(CN)C=CC(=C1)C(F)(F)F

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results