accessibility menu, dialog, popup

UserName

CAS RN 7705-07-9

ClClClTi

CAS RN 7705-07-9

IUPAC Name: titanium(4+) tetrachloride
Synonyms: titanium tetrachloride
Molecular Weight (g/mol): 189.67
Molecular Formula: Cl4Ti
InChi Key: XJDNKRIXUMDJCW-UHFFFAOYSA-J
SMILES: [Cl-].[Cl-].[Cl-].[Cl-].[Ti+4]
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
Percent Purity 
  • (4)
pH 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (4)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (4)
  • (4)
  • (2)