accessibility menu, dialog, popup

UserName

H2NH2NNSSNH2NSNH2OON(E/Z)

CAS RN 76824-35-6

IUPAC Name: 3-[({2-[(diaminomethylidene)amino]-1,3-thiazol-4-yl}methyl)sulfanyl]-N'-sulfamoylpropanimidamide
Synonyms: famotidine famotidina pepcid famotidinum 法莫替丁
Molecular Weight (g/mol): 337.44
Molecular Formula: C8H15N7O2S3
InChi Key: XUFQPHANEAPEMJ-UHFFFAOYSA-N
SMILES: NC(N)=NC1=NC(CSCCC(N)=NS(N)(=O)=O)=CS1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (1)
  • (1)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results