accessibility menu, dialog, popup

UserName

CAS RN 7682-20-4

H3CONH2H2NHCl

CAS RN 7682-20-4

IUPAC Name: 2-aminobutanamide hydrochloride
Synonyms: 2-aminobutanamide hydrochloride
Molecular Weight (g/mol): 138.6
Molecular Formula: C4H11ClN2O
InChi Key: HDBMIDJFXOYCGK-UHFFFAOYNA-N
SMILES: Cl.CCC(N)C(N)=O
Molecular Weight (g/mol) 
  • (3)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results