accessibility menu, dialog, popup

UserName

CAS RN 7681-65-4

IUPAC Name: λ¹-copper(1+) iodide
Synonyms: cuprous iodide
Molecular Weight (g/mol): 190.45
Molecular Formula: CuI
InChi Key: LSXDOTMGLUJQCM-UHFFFAOYSA-M
SMILES: [Cu+].[I-]
Quantity 
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
Physical Form 
  • (3)
  • (8)
  • (1)
Percent Purity 
  • (3)
  • (8)
Grade 
  • (3)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (3)
  • (3)
  • (2)
  • (3)
  • (2)
  • (1)
  • (1)
  • (3)
  • (8)
  • (1)
  • (3)
  • (8)
  • (3)