CAS RN 75847-73-3
CAS RN 75847-73-3
IUPAC Name:
(2S)-1-[(2S)-2-{[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino}propanoyl]pyrrolidine-2-carboxylic acid
Synonyms:
enalapril enalaprila
Molecular Weight (g/mol):
376.45
Molecular Formula:
C20H28N2O5
InChi Key:
GBXSMTUPTTWBMN-XIRDDKMYSA-N
SMILES:
CCOC(=O)[C@H](CCC1=CC=CC=C1)N[C@@H](C)C(=O)N1CCC[C@H]1C(O)=O
1
–
1
of
1
results
1
Enalapril, Thermo Scientific™
CAS: 75847-73-3 Molecular Formula: C20H28N2O5 Molecular Weight (g/mol): 376.453 MDL Number: MFCD00865774 InChI Key: GBXSMTUPTTWBMN-XIRDDKMYSA-N Synonym: enalapril,enalaprilum,vasotec,enalaprila,enalapril tn,unii-69pn84io1a,enalapril inn,enalapril richet,chembl578,enalaprilum inn-latin PubChem CID: 5388962 ChEBI: CHEBI:4784 IUPAC Name: (2S)-1-[(2S)-2-[[(2S)-1-ethoxy-1-oxo-4-phenylbutan-2-yl]amino]propanoyl]pyrrolidine-2-carboxylic acid SMILES: CCOC(=O)C(CCC1=CC=CC=C1)NC(C)C(=O)N2CCCC2C(=O)O