accessibility menu, dialog, popup

UserName

H3CNHONHOHOHCH3OOHOOH(E/Z)

CAS RN 753498-25-8

IUPAC Name: 5-{2-[(5,6-diethyl-2,3-dihydro-1H-inden-2-yl)amino]-1-hydroxyethyl}-8-hydroxy-1,2-dihydroquinolin-2-one; but-2-enedioic acid
Synonyms: butenedioic acid; indacaterol
Molecular Weight (g/mol): 508.57
Molecular Formula: C28H32N2O7
InChi Key: IREJFXIHXRZFER-UHFFFAOYNA-N
SMILES: OC(=O)C=CC(O)=O.CCC1=C(CC)C=C2CC(CC2=C1)NCC(O)C1=C2C=CC(=O)NC2=C(O)C=C1
Molecular Weight (g/mol) 
  • (4)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results