CAS RN 7517-19-3
CAS RN 7517-19-3
IUPAC Name:
hydrogen methyl 2-amino-4-methylpentanoate chloride
Synonyms:
hydrogen methyl 2-amino-4-methylpentanoate chloride
Molecular Weight (g/mol):
181.66
Molecular Formula:
C7H16ClNO2
InChi Key:
DODCBMODXGJOKD-UHFFFAOYNA-N
SMILES:
[H+].[Cl-].COC(=O)C(N)CC(C)C
1
–
2
of
2
results
1
Promotions Available
Thermo Scientific Chemicals L-Leucine methyl ester hydrochloride, 98%
Thermo Scientific Chemicals L-Leucine methyl ester hydrochloride, 98%
2
L-Leucine methyl ester hydrochloride, 98+%, Thermo Scientific Chemicals
CAS: 7517-19-3 Molecular Formula: C7H16ClNO2 Molecular Weight (g/mol): 181.66 MDL Number: MFCD00012494 InChI Key: DODCBMODXGJOKD-HQFIOVGONA-N Synonym: l-leucine methyl ester hydrochloride,h-leu-ome.hcl,methyl l-leucinate hydrochloride,s-methyl 2-amino-4-methylpentanoate hydrochloride,h-leu-ome. hcl,leucine methyl ester hydrochloride,h-leu-ome hydrochloride,l-leucine methyl ester hydrochloride;h-leu-ome.hcl,s-2-amino-4-methyl-pentanoic acid methyl ester hydrochloride,l-leucine, methyl ester, monohydrochloride PubChem CID: 2723695 IUPAC Name: methyl (2S)-2-amino-4-methylpentanoate;hydrochloride SMILES: Cl.COC(=O)[C@@H](N)CC(C)C